COMH42116CDA Combi-Blocks, Inc.

4,4,5,5-tetramethyl-2-[6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl]-1,3,2-dioxaborolane

  • MDL number MFCD22381081
  • Cas Number : 849543-98-2
  • Molecular Weight : 380.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1413
Availability details for
Quantity
  • Availability
  • Add to Cart