COMH42116CDB Combi-Blocks, Inc.

Methyl 5-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD14706694
  • Cas Number : 1088994-18-6
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1585
Availability details for
Quantity
  • Availability
  • Add to Cart