COMH42116CDD Combi-Blocks, Inc.

2-(3,5-difluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22414481
  • Cas Number : 890839-37-9
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1607
Availability details for
Quantity
  • Availability
  • Add to Cart