COMH42116CDF Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylic acid

  • MDL number MFCD24039389
  • Cas Number : 1426082-97-4
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1655
Availability details for
Quantity
  • Availability
  • Add to Cart