COMH42116CE2 Combi-Blocks, Inc.

3-Methoxy-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD23380405
  • Cas Number : 1374325-17-3
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1802

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1802
Availability details for
Quantity
  • Availability
  • Add to Cart