COMH42116CE8 Combi-Blocks, Inc.

1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbonyl}piperidin-4-ol

  • MDL number MFCD10698502
  • Cas Number : 1100094-82-3
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2899
Availability details for
Quantity
  • Availability
  • Add to Cart