COMH42116CE9 Combi-Blocks, Inc.

1-[3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1h-pyrazole

  • MDL number MFCD11043448
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2937
Availability details for
Quantity
  • Availability
  • Add to Cart