COMH42116CEB Combi-Blocks, Inc.

N-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinamide

  • MDL number MFCD11878277
  • Cas Number : 1201644-52-1
  • Molecular Weight : 288.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinamide
Inchi Code 1S/C15H21BN2O3/c1-14(2)15(3,4)21-16(20-14)11-7-10(8-17-9-11)13(19)18-12-5-6-12/h7-9,12H,5-6H2,1-4H3,(H,18,19)
Inchi Key MJGNAJWFUMNJLT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3262
Availability details for
Quantity
  • Availability
  • Add to Cart