COMH42116CED Combi-Blocks, Inc.

2-(3-Trifluoromethylphenyl)thiazole-4-boronic acid pinacol ester

  • MDL number MFCD11869354
  • Cas Number : 1402237-78-8
  • Molecular Weight : 355.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3473
Availability details for
Quantity
  • Availability
  • Add to Cart