COMH42116CEF Combi-Blocks, Inc.

2-(Cyclohexyl)thiazole-4-boronic acid pinacol ester

  • MDL number MFCD11223319
  • Cas Number : 1402166-61-3
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3580
Availability details for
Quantity
  • Availability
  • Add to Cart