COMH42116CF9 Combi-Blocks, Inc.

tert-Butyl 4-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxymethyl]piperidine-1-carboxylate

  • MDL number MFCD12827857
  • Cas Number : 1310383-45-9
  • Molecular Weight : 417.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6523
Availability details for
Quantity
  • Availability
  • Add to Cart