COMH42116CFA Combi-Blocks, Inc.

N-[2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-pyridinyl]-cyclopropanesulfonamide

  • MDL number MFCD12923394
  • Cas Number : 1083326-71-9
  • Molecular Weight : 354.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6583
Availability details for
Quantity
  • Availability
  • Add to Cart