COMH42116CFF Combi-Blocks, Inc.

Methyl 3-amino-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine-2-carboxylate

  • MDL number MFCD16036084
  • Cas Number : 1874279-25-0
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7443
Availability details for
Quantity
  • Availability
  • Add to Cart