COMH42116D0D Combi-Blocks, Inc.

tert-Butyl 4-formyloxazol-2-ylcarbamate

  • MDL number MFCD13189393
  • Cas Number : 1060816-37-6
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0030
Availability details for
Quantity
  • Availability
  • Add to Cart