COMH42116D12 Combi-Blocks, Inc.

Amino-peg23-amine

  • MDL number MFCD31580136
  • Molecular Weight : 1073.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C48H100N2O23/c49-1-3-51-5-7-53-9-11-55-13-15-57-17-19-59-21-23-61-25-27-63-29-31-65-33-35-67-37-39-69-41-43-71-45-47-73-48-46-72-44-42-70-40-38-68-36-34-66-32-30-64-28-26-62-24-22-60-20-18-58-16-14-56-12-10-54-8-6-52-4-2-50/h1-50H2
Inchi Key JXEMKMYTIDWELB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0041
Availability details for
Quantity
  • Availability
  • Add to Cart