COMH42116D17 Combi-Blocks, Inc.

5-Aminopyrazolo[1,5-a]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD31619053
  • Cas Number : 2279124-20-6
  • Molecular Weight : 213.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0066
Availability details for
Quantity
  • Availability
  • Add to Cart