COMH42116D22 Combi-Blocks, Inc.

5-[(Cyclobutylmethyl)sulfanyl]-1,3,4-thiadiazol-2-amine

  • MDL number MFCD21240145
  • Cas Number : 1467128-54-6
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0134
Availability details for
Quantity
  • Availability
  • Add to Cart