COMH42116D28 Combi-Blocks, Inc.

M-Peg9-2-methylacrylate

  • MDL number MFCD30828683
  • Cas Number : 35236-69-2
  • Molecular Weight : 496.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-yl methacrylate
Inchi Code 1S/C23H44O11/c1-22(2)23(24)34-21-20-33-19-18-32-17-16-31-15-14-30-13-12-29-11-10-28-9-8-27-7-6-26-5-4-25-3/h1,4-21H2,2-3H3
Inchi Key TXHZNLCKXHJYNX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0166
Availability details for
Quantity
  • Availability
  • Add to Cart