COMH42116D36 Combi-Blocks, Inc.

Ethyl 2-amino-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD31581336
  • Cas Number : 2270908-42-2
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0225
Availability details for
Quantity
  • Availability
  • Add to Cart