COMH42116D41 Combi-Blocks, Inc.

4,4-Diethylpyrrolidin-2-one

  • MDL number MFCD15146419
  • Cas Number : 858450-14-3
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0349
Availability details for
Quantity
  • Availability
  • Add to Cart