COMH42116D52 Combi-Blocks, Inc.

6-(2-Amino-5-bromophenyl)-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2h)-one

  • MDL number MFCD00455171
  • Cas Number : 129951-97-9
  • Molecular Weight : 299.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0433
Availability details for
Quantity
  • Availability
  • Add to Cart