COMH42116D55 Combi-Blocks, Inc.

1H-Phenalene-1,2,3-trione

  • MDL number MFCD00453272
  • Cas Number : 5116-63-2
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0452
Availability details for
Quantity
  • Availability
  • Add to Cart