COMH42116D62 Combi-Blocks, Inc.

Novobiocin sodium salt

  • MDL number MFCD30207859
  • Cas Number : 1476-53-5
  • Molecular Weight : 634.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0497
Availability details for
Quantity
  • Availability
  • Add to Cart