COMH42116D6B Combi-Blocks, Inc.

2-Cyano-3-methyl-pent-2-enedioic acid 1-ethyl ester 5-methyl ester

  • MDL number MFCD22385532
  • Cas Number : 1366392-10-0
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl 5-methyl (2E)-2-cyano-3-methyl-2-pentenedioate
Inchi Code 1S/C10H13NO4/c1-4-15-10(13)8(6-11)7(2)5-9(12)14-3/h4-5H2,1-3H3/b8-7+
Inchi Key LGPULUNQERJYGI-BQYQJAHWSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0533
Availability details for
Quantity
  • Availability
  • Add to Cart