COMH42116D78 Combi-Blocks, Inc.

Methyl 2-[(4-fluoro-3-methoxyphenyl)(hydroxy)methyl]acrylate

  • MDL number MFCD31583410
  • Cas Number : 2270909-57-2
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-((4-fluoro-3-methoxyphenyl)(hydroxy)methyl)acrylate
Inchi Code 1S/C12H13FO4/c1-7(12(15)17-3)11(14)8-4-5-9(13)10(6-8)16-2/h4-6,11,14H,1H2,2-3H3
Inchi Key DQOTWQDPTIRPCB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0615

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0615
Availability details for
Quantity
  • Availability
  • Add to Cart