COMH42116D7A Combi-Blocks, Inc.

Fmoc-aminooxy-peg12-nhs ester

  • MDL number MFCD31615435
  • Molecular Weight : 953.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0624
Availability details for
Quantity
  • Availability
  • Add to Cart