COMH42116D7B Combi-Blocks, Inc.

Methyl 5-(4-fluoro-3-methoxyphenyl)-3-nitroisoxazole-4-carboxylate

  • MDL number MFCD31583412
  • Cas Number : 2270905-20-7
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0626
Availability details for
Quantity
  • Availability
  • Add to Cart