COMH42116D7D Combi-Blocks, Inc.

N-(4-Bromophenyl)-4-(3,5-dimethyl-1h-pyrazol-1-yl)-6-hydrazino-1,3,5-triazin-2-amine

  • MDL number MFCD02255691
  • Cas Number : 1158450-00-0
  • Molecular Weight : 375.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0635
Availability details for
Quantity
  • Availability
  • Add to Cart