COMH42116D7F Combi-Blocks, Inc.

2,5,8,11-Tetraoxatetradecan-14-ol

  • MDL number MFCD26911025
  • Cas Number : 145526-76-7
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0645
Availability details for
Quantity
  • Availability
  • Add to Cart