COMH42116D81 Combi-Blocks, Inc.

4-Formyl-2,5-dimethyl-1-(3-methylbenzyl)-1h-pyrrole-3-carboxylic acid

  • MDL number MFCD14281684
  • Cas Number : 1172712-44-5
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0653
Availability details for
Quantity
  • Availability
  • Add to Cart