COMH42116D82 Combi-Blocks, Inc.

N-[(1-Ethyl-5-methyl-1h-pyrazol-4-yl)methyl]ethanamine

  • MDL number MFCD08685650
  • Cas Number : 956935-38-9
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-N-[(1-ethyl-5-methyl-1H-pyrazol-4-yl)methyl]amine
Inchi Code 1S/C9H17N3/c1-4-10-6-9-7-11-12(5-2)8(9)3/h7,10H,4-6H2,1-3H3
Inchi Key OHGSEVDQYHENGZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0656
Availability details for
Quantity
  • Availability
  • Add to Cart