COMH42116D88 Combi-Blocks, Inc.

7-Fluoro-6-hydroxybenzofuran

  • MDL number MFCD31558574
  • Cas Number : 1429789-84-3
  • Molecular Weight : 152.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0691
Availability details for
Quantity
  • Availability
  • Add to Cart