COMH42116D97 Combi-Blocks, Inc.

(4-Methoxy-3,5-dimethylpyridin-2-yl)acetonitrile

  • MDL number MFCD11131917
  • Cas Number : 218921-06-3
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0739
Availability details for
Quantity
  • Availability
  • Add to Cart