COMH42116D9E Combi-Blocks, Inc.

(3S,3Ar,5r,6ar,7r)-6-iodo-2-oxooctahydro-3,5-methanocyclopenta[b]pyrrole-7-carboxylic acid

  • MDL number MFCD31613612
  • Cas Number : 2365398-43-0
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0757
Availability details for
Quantity
  • Availability
  • Add to Cart