COMH42116DA0 Combi-Blocks, Inc.

N-Dbco-n-bis(peg2-nhs ester)

  • MDL number MFCD29912779
  • Cas Number : 2128735-29-3
  • Molecular Weight : 818.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0763
Availability details for
Quantity
  • Availability
  • Add to Cart