COMH42116DA2 Combi-Blocks, Inc.

(1,3-Bis(diphenylphosphino)propane)palladium(ii) chloride dichloromethane adduct

  • MDL number MFCD28139116
  • Molecular Weight : 674.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0766
Availability details for
Quantity
  • Availability
  • Add to Cart