COMH42116DA5 Combi-Blocks, Inc.

2,2,3,3-Tetrafluorocyclobutanecarbonitrile

  • MDL number MFCD00039434
  • Cas Number : 356-81-0
  • Molecular Weight : 153.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0786
Availability details for
Quantity
  • Availability
  • Add to Cart