COMH42116DA8 Combi-Blocks, Inc.

Methyl 2,5-dimethyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD31581338
  • Cas Number : 2270905-24-1
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methyl-5-methylene-4,5-dihydro-[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Inchi Code 1S/C9H10N4O2/c1-5-7(8(14)15-3)4-13-9(10-5)11-6(2)12-13/h4H,1H2,2-3H3,(H,10,11,12)
Inchi Key YUCIZFMFRXVZFB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0796
Availability details for
Quantity
  • Availability
  • Add to Cart