COMH42116DA9 Combi-Blocks, Inc.

3-[(3-Methylbutyl)thio]-1h-1,2,4-triazol-5-amine

  • MDL number MFCD08592473
  • Cas Number : 861925-01-1
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0798
Availability details for
Quantity
  • Availability
  • Add to Cart