COMH42116DAA Combi-Blocks, Inc.

6,6-Dimethyl-1,4-dioxane-2-carboxylic acid

  • MDL number MFCD31583462
  • Cas Number : 2168882-32-2
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0801
Availability details for
Quantity
  • Availability
  • Add to Cart