COMH42116DAB Combi-Blocks, Inc.

[2-(1H-Imidazol-1-yl)phenyl]amine dihydrochloride

  • MDL number MFCD09759124
  • Cas Number : 1211477-87-0
  • Molecular Weight : 232.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-imidazol-1-yl)aniline dihydrochloride
Inchi Code 1S/C9H9N3.2ClH/c10-8-3-1-2-4-9(8)12-6-5-11-7-12;;/h1-7H,10H2;2*1H
Inchi Key GMANGTCLSBAIKQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0805
Availability details for
Quantity
  • Availability
  • Add to Cart