COMH42116DAC Combi-Blocks, Inc.

tert-Butyl n-[(1s)-1-(hydrazinecarbonyl)-3-methylbutyl]carbamate

  • MDL number MFCD11108873
  • Cas Number : 77284-59-4
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0810
Availability details for
Quantity
  • Availability
  • Add to Cart