COMH42116DBA Combi-Blocks, Inc.

[2-(1-Adamantyloxy)propyl]amine hydrochloride

  • MDL number MFCD27756544
  • Cas Number : 21623-91-6
  • Molecular Weight : 245.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0867
Availability details for
Quantity
  • Availability
  • Add to Cart