COMH42116DBC Combi-Blocks, Inc.

3-(Dimethylamino)-5-(4-fluoro-3-methoxyphenyl)isoxazole-4-carboxylic acid

  • MDL number MFCD31583414
  • Cas Number : 2270912-39-3
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0877
Availability details for
Quantity
  • Availability
  • Add to Cart