COMH42116DC0 Combi-Blocks, Inc.

N,N-Dibutylhydrazinecarbothioamide hydrochloride

  • MDL number MFCD00463361
  • Cas Number : 6499-16-7
  • Molecular Weight : 239.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N-dibutylhydrazinecarbothioamide hydrochloride
Inchi Code 1S/C9H21N3S.ClH/c1-3-5-7-12(8-6-4-2)9(13)11-10;/h3-8,10H2,1-2H3,(H,11,13);1H
Inchi Key DHOKEGWISNXRTD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0892
Availability details for
Quantity
  • Availability
  • Add to Cart