COMH42116DCE Combi-Blocks, Inc.

Potassium (5-amino-3-bromo-1h-1,2,4-triazol-1-yl)acetate

  • MDL number MFCD31583432
  • Cas Number : 2270909-67-4
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0973
Availability details for
Quantity
  • Availability
  • Add to Cart