COMH42116DD1 Combi-Blocks, Inc.

2-Methoxy-3-nitro-5-(trifluoromethoxy)benzoic acid

  • MDL number MFCD31583448
  • Cas Number : 2270906-37-9
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0986
Availability details for
Quantity
  • Availability
  • Add to Cart