COMH42116DD3 Combi-Blocks, Inc.

3-[2,5-Dioxo-1-(2-phenylethyl)imidazolidin-4-yl]propanoic acid

  • MDL number MFCD18485566
  • Cas Number : 1214818-05-9
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[2,5-dioxo-1-(2-phenylethyl)-4-imidazolidinyl]propanoic acid
Inchi Code 1S/C14H16N2O4/c17-12(18)7-6-11-13(19)16(14(20)15-11)9-8-10-4-2-1-3-5-10/h1-5,11H,6-9H2,(H,15,20)(H,17,18)
Inchi Key HUYGOICTAVMQIF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0995
Availability details for
Quantity
  • Availability
  • Add to Cart