COMH42116DD8 Combi-Blocks, Inc.

6-(1H-Pyrazol-1-yl)hexan-3-one

  • MDL number MFCD31557144
  • Cas Number : 1692572-10-3
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1013
Availability details for
Quantity
  • Availability
  • Add to Cart