COMH42116DD9 Combi-Blocks, Inc.

N,N-Bis(2-cyanoethyl)formamide

  • MDL number MFCD00040894
  • Cas Number : 3445-84-9
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1014
Availability details for
Quantity
  • Availability
  • Add to Cart